Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-[(3R)-1-methyl-2-oxo-5-phenyl-3H-1,4-benzodiazepin-3-yl]-3-(3-methylphenyl)urea | 118101-09-0 | MFCD00869756 | 99.9% | 398.46 | C24H22N4O2 | 1 ML
SDP

Catalog No. 5000241115
Encompass_Preferred

L-365260 is a selective, orally active antagonist of the gastrin/cholecystokinin B (CCK-B/CCK2) receptor used in pharmacology and receptor biology research. It is supplied in high-purity solid form and can be provided as concentrated solution formats for in vitro and in vivo laboratory studies.

  • High purity (99.85% by HPLC).
  • Molecular formula C24H22N4O2.
  • Molecular weight 398.46 g/mol.
  • Appearance white to off-white solid.
  • Storage: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C (up to 6 months).
  • Available as a 10 mM solution in 1 mL volumes for experimental use.

Catalog No. 50-002-41115 Supplier Medchemexpress LLC Supplier No. HY10684010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-365260 is a selective, orally active antagonist of the gastrin/cholecystokinin B (CCK-B/CCK2) receptor used in pharmacology and receptor biology research. It is supplied in high-purity solid form and can be provided as concentrated solution formats for in vitro and in vivo laboratory studies.

  • High purity (99.85% by HPLC).
  • Molecular formula C24H22N4O2.
  • Molecular weight 398.46 g/mol.
  • Appearance white to off-white solid.
  • Storage: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C (up to 6 months).
  • Available as a 10 mM solution in 1 mL volumes for experimental use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.