Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[4-(2-Phenylethyl)benzyl]naphthalene 98.0+%, TCI America™
SDP

Catalog No. P11811G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11811G 1 g
P118125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11811G Supplier TCI America Supplier No. P11811G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 127833-53-8
Molecular Formula C25H22
Molecular Weight (g/mol) 322.451
MDL Number MFCD00191572
InChI Key UKRRCRMJNMWWMZ-UHFFFAOYSA-N
Synonym 1-[4-(2-Phenethyl)benzyl]naphthalene
PubChem CID 14833659
IUPAC Name 1-[[4-(2-phenylethyl)phenyl]methyl]naphthalene
SMILES C1=CC=C(C=C1)CCC2=CC=C(C=C2)CC3=CC=CC4=CC=CC=C43

Specifications

Melting Point 66°C
Color White-Yellow
Quantity 1 g
Formula Weight 322.45
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-[4-(2-Phenylethyl)benzyl]naphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.