Learn More
Chemglass Life Sciences Replacement Screw Only, 1/4 - 20 x 1/2" slotted fillister head 20% glass filled
SDP
Catalog No. 501531444
Encompass_Preferred
Replacement Screw Only, 1/4 - 20 x 1/2" slotted fillister head 20% glass filled. Replacement screw for CG-2095 series of PTFE agitators. Pictured are fully assembled agitators. THIS ITEM IS SCREW ONLY!
Catalog No. 50-153-1444 Supplier Chemglass Life Sciences Supplier No. CG209599
Replacement Screw Only, 1/4 - 20 x 1/2" slotted fillister head 20% glass filled. Replacement screw for CG-2095 series of PTFE agitators. Pictured are fully assembled agitators. THIS ITEM IS SCREW ONLY!