Learn More
IDEX Health & Science Upchurch Flangeless Fitting Natural, PEEK, 1/4-28 Flat-Bottom, for 1/16" OD
Supplier:  IDEX Health & Science Upchurch XP230
Encompass_Preferred
This item has a minimum qty of 25 per supplier requirements.Our Flangeless PEEK Fittings are designed to include both a nut and ferrule for your convenience. Flangeless fitting are dependable, easy to use and easy to replace. Manufactured with biocompatible PEEK it offers excellent chemical resistance. It has a 1/4 Flat-Bottom port configuration and is for 1/16" OD tubing. Our Fitting System comes with a P-200 ferrule.
Catalog No. NC2136107