Learn More
Ace Glass, Inc. 1/4-28 female to 1/4-28 female peek union.
SDP
Catalog No. 50972360
Encompass
-28 female to -28 female PEEK union. A chemically inert connector for connecting PTFE or Tefzel tubing to Chromatography adapters.
Catalog No. 50-972-360 Supplier Ace Glass, Inc. Supplier No. 585440
-28 female to -28 female PEEK union. A chemically inert connector for connecting PTFE or Tefzel tubing to Chromatography adapters.