Learn More
Medchemexpress LLC 1-[4-[(2S)-2-(morpholin-4-ylmethyl)pyrrolidin-1-yl]sulfonylphenyl]-3-[4-(trifluoromethyl)phenyl]urea | 2230496-99-6 | 99.9% | 512.55 | C23H27F3N4O4S | 5 MG
SDP
Catalog No. 5000221409
Encompass_Preferred
ZL0590 is a potent, orally active small-molecule inhibitor selective for the BRD4 BD1 bromodomain. It has an IC50 of approximately 90 nM against human BRD4 BD1 and demonstrates anti-inflammatory efficacy in preclinical models, making it suitable for research into bromodomain biology and inflammatory disease mechanisms.
  • Potent BRD4 BD1 inhibition (IC50 ≈ 90 nM)
  • Selective for BD1 over other bromodomains
  • Demonstrated anti-inflammatory activity in animal models
  • Appropriate for in vitro and in vivo research applications
  • Soluble in DMSO for solution preparation
  • High chemical purity (~99.9%)
Catalog No. 50-002-21409 Supplier Medchemexpress LLC Supplier No. HY1453105MG
ZL0590 is a potent, orally active small-molecule inhibitor selective for the BRD4 BD1 bromodomain. It has an IC50 of approximately 90 nM against human BRD4 BD1 and demonstrates anti-inflammatory efficacy in preclinical models, making it suitable for research into bromodomain biology and inflammatory disease mechanisms.
  • Potent BRD4 BD1 inhibition (IC50 ≈ 90 nM)
  • Selective for BD1 over other bromodomains
  • Demonstrated anti-inflammatory activity in animal models
  • Appropriate for in vitro and in vivo research applications
  • Soluble in DMSO for solution preparation
  • High chemical purity (~99.9%)