Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-[4-(3,5-dichlorophenyl)-3-fluorophenyl]cyclopropane-1-carboxylic acid | 876625-29-5 | 99.5% | 325.16 g·mol⁻¹ | C16H11Cl2FO2 | 10 MG
SDP

Catalog No. 5000287225
Encompass_Preferred

This compound is a research-grade small-molecule inhibitor of aldo-keto reductase 1C3 (AKR1C3) that selectively blocks AKR1C3-mediated reduction of doxorubicin and can reverse doxorubicin resistance in AKR1C3-overexpressing cells. Supplied as a high-purity solid, it is intended for biochemical and cellular research applications.

  • Target: AKR1C3 inhibitor.
  • Mechanism: selectively blocks AKR1C3-mediated reduction of doxorubicin.
  • Purity: 99.5% (reported value).
  • Physical form: white to off-white solid.
  • Solubility: soluble in DMSO (≈100 mg/mL); in vivo formulations reported in DMSO/corn oil.
  • Storage: powder stable at -20°C for long-term storage; solutions require low-temperature storage.

Catalog No. 50-002-87225 Supplier Medchemexpress LLC Supplier No. HY15503510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This compound is a research-grade small-molecule inhibitor of aldo-keto reductase 1C3 (AKR1C3) that selectively blocks AKR1C3-mediated reduction of doxorubicin and can reverse doxorubicin resistance in AKR1C3-overexpressing cells. Supplied as a high-purity solid, it is intended for biochemical and cellular research applications.

  • Target: AKR1C3 inhibitor.
  • Mechanism: selectively blocks AKR1C3-mediated reduction of doxorubicin.
  • Purity: 99.5% (reported value).
  • Physical form: white to off-white solid.
  • Solubility: soluble in DMSO (≈100 mg/mL); in vivo formulations reported in DMSO/corn oil.
  • Storage: powder stable at -20°C for long-term storage; solutions require low-temperature storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.