Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-[4-(3-tert-butyl-4-hydroxyphenoxy)phenyl]ethan-1-one | 2735803-28-6 | MFCD34676378 | 99.2% | C18H20O3 | 10MG
SDP

Catalog No. 5000260093
Encompass_Preferred

DS45500853 is a small-molecule agonist of estrogen-related receptor α (ERRα) supplied as a high-purity solid for research use. It is characterized by formula C18H20O3 and molecular weight 284.35 g·mol-1, and is intended for biochemical and cell-based assays targeting ERRα.

  • High purity: 99.2%.
  • Target activity: estrogen-related receptor α (ERRα) agonist.
  • Physical form: solid, light yellow to yellow.
  • Molecular data: C18H20O3, 284.35 g·mol-1.
  • Storage stability: powder stable at -20°C for years; in solvent stable at -80°C for months.
  • Solubility: soluble in DMSO and ethanol.

Catalog No. 50-002-60093 Supplier Medchemexpress LLC Supplier No. HY13220510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DS45500853 is a small-molecule agonist of estrogen-related receptor α (ERRα) supplied as a high-purity solid for research use. It is characterized by formula C18H20O3 and molecular weight 284.35 g·mol-1, and is intended for biochemical and cell-based assays targeting ERRα.

  • High purity: 99.2%.
  • Target activity: estrogen-related receptor α (ERRα) agonist.
  • Physical form: solid, light yellow to yellow.
  • Molecular data: C18H20O3, 284.35 g·mol-1.
  • Storage stability: powder stable at -20°C for years; in solvent stable at -80°C for months.
  • Solubility: soluble in DMSO and ethanol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.