Learn More
Medchemexpress LLC 1-[[4-[4-[[4-(4-chloro-N-methylanilino)quinolin-1-ium-1-yl]methyl]phenyl]phenyl]methyl]-N-(4-chlor... | 850807-63-5 | 98.4% | 877.53 g/mol | C46H38Br2Cl2N4 | 5 MG
SDP
Catalog No. 5000211949
Encompass_Preferred
RSM-932A is a small-molecule choline kinase α (ChoKα) inhibitor used as a research tool in preclinical oncology studies. It shows potent antiproliferative activity in vitro and antitumor efficacy in vivo, and is commonly used to study ChoKα biology and evaluate therapeutic hypotheses in cellular and animal models.
  • Selective ChoKα inhibitor with reported IC50 ≈ 1 μM and ~33 μM for ChoKβ.
  • Potent antiproliferative activity across tumor-derived cell lines.
  • Demonstrated antitumor efficacy in xenograft models.
  • High-purity synthetic compound suitable for research applications.
  • Solid form; molecular weight 877.53 g/mol.
  • Store sealed at 4°C; long-term storage in solution recommended at -80°C.
Catalog No. 50-002-11949 Supplier Medchemexpress LLC Supplier No. HY1011445MG
RSM-932A is a small-molecule choline kinase α (ChoKα) inhibitor used as a research tool in preclinical oncology studies. It shows potent antiproliferative activity in vitro and antitumor efficacy in vivo, and is commonly used to study ChoKα biology and evaluate therapeutic hypotheses in cellular and animal models.
  • Selective ChoKα inhibitor with reported IC50 ≈ 1 μM and ~33 μM for ChoKβ.
  • Potent antiproliferative activity across tumor-derived cell lines.
  • Demonstrated antitumor efficacy in xenograft models.
  • High-purity synthetic compound suitable for research applications.
  • Solid form; molecular weight 877.53 g/mol.
  • Store sealed at 4°C; long-term storage in solution recommended at -80°C.