Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-[[4-[4-[[4-(4-chloro-N-methylanilino)quinolin-1-ium-1-yl]methyl]phenyl]phenyl]methyl]-N-(4-chlorop | 850807-63-5 | 98.4% | 877.53 | C46H38Br2Cl2N4 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY10114450MG

Encompass_Preferred

Selective choline kinase alpha (ChoKα) inhibitor for biochemical and cell-based cancer research. It demonstrates potent in vitro anti-proliferative activity and in vivo antitumor efficacy in xenograft models. Supplied as a solid for laboratory use in small-scale quantities.

  • Selective ChoKα inhibition (IC50 ≈ 1 μM).
  • Activity against multiple tumor-derived cell lines (IC50 ~1.3-7.1 μM).
  • High purity (98.4%).
  • Molecular weight 877.53; CAS 850807-63-5.
  • Storage: 4 °C sealed; in solvent -80 °C (6 months), -20 °C (1 month).

Catalog No. 50-002-13524


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.