Learn More
eMolecules​ 1227068-67-8 | 1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridin-1(2H)-yl)ethanone | MFCD18427628 | 1g
Supplier:  eMolecules​ 525035441
Encompass_Preferred
Ambeed | (3aR3aR8aS8aS)-88a88a-Tetrahydro-3aH3aH-22-biindeno[12-d]oxazole | 100mg | 633660659 | A1378974 | 1354558-61-4 | 316.360 | C20H16N2O2
If you are unable to find the chemical you are looking for, make sure you are logged into your fishersci.com account and click on the following link:
eMolecules Building Block Tool<|a>
Catalog No. 50-213-8638