Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4,5,8-Naphthalenetetracarboxdiimide 95.0+%, TCI America™
SDP

Catalog No. N05361G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05361G 1 g
N05365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N05361G Supplier TCI America Supplier No. N05361G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5690-24-4
Molecular Formula C14H6N2O4
Molecular Weight (g/mol) 266.21
MDL Number MFCD00047649
InChI Key BODUWJSFPLUDMP-UHFFFAOYSA-N
PubChem CID 79771
IUPAC Name 6,13-diazatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1,3,8,10,15-pentaene-5,7,12,14-tetrone
SMILES O=C1NC(=O)C2=CC=C3C(=O)NC(=O)C4=CC=C1C2=C34

Specifications

Quantity 1 g
Formula Weight 266.21
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,4,5,8-Naphthalenetetracarboxdiimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.