Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4,5,8-Naphthalenetetracarboxylic Acid (contains Monoanhydride) 60.0+%, TCI America™
SDP

Catalog No. N077025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N077025G 25 g
N0770250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N077025G Supplier TCI America Supplier No. N077025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 128-97-2
Molecular Formula C14H8O8
Molecular Weight (g/mol) 304.21
MDL Number MFCD00004012
InChI Key OLAPPGSPBNVTRF-UHFFFAOYSA-N
PubChem CID 31422
IUPAC Name naphthalene-1,4,5,8-tetracarboxylic acid
SMILES OC(=O)C1=CC=C(C(O)=O)C2=C(C=CC(C(O)=O)=C12)C(O)=O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 304.21
Percent Purity ≥60.0% (NMR)
Physical Form Crystalline Powder
Chemical Name or Material 1,4,5,8-Naphthalenetetracarboxylic Acid (contains Monoanhydride)
EINECSNumber (4)-1009
RTECSNumber QK3690000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.