Learn More
Medchemexpress LLC 1-[4-[5-carbamoyl-2-[(2-ethyl-5-methylpyrazole-3-carbonyl)amino]benzimidazol-1-yl]butyl]-2-[(2-ethyl... | 2138300-40-8 | 99.5% | 678.74 g·mol⁻¹ | C34H38N12O4 | 1MG
SDP
Catalog No. 5000235629
Encompass_Preferred
STING agonist-4 is a small-molecule agonist of the stimulator of interferon genes (STING) receptor used as a research reagent to modulate innate immune signaling. It exhibits nanomolar potency and is typically used in in vitro pathway activation studies and preclinical immunology research.
  • Potent STING agonist with IC50 ≈ 20 nM.
  • Chemical formula C34H38N12O4 and molecular weight 678.74 g·mol⁻¹.
  • High reported purity suitable for research use (≈99.5%).
  • Offered in small pack sizes for experimental flexibility.
  • Applicable for in vitro immune signaling and preclinical studies.
Catalog No. 50-002-35629 Supplier Medchemexpress LLC Supplier No. HY1239431MG
STING agonist-4 is a small-molecule agonist of the stimulator of interferon genes (STING) receptor used as a research reagent to modulate innate immune signaling. It exhibits nanomolar potency and is typically used in in vitro pathway activation studies and preclinical immunology research.
  • Potent STING agonist with IC50 ≈ 20 nM.
  • Chemical formula C34H38N12O4 and molecular weight 678.74 g·mol⁻¹.
  • High reported purity suitable for research use (≈99.5%).
  • Offered in small pack sizes for experimental flexibility.
  • Applicable for in vitro immune signaling and preclinical studies.