Learn More
Medchemexpress LLC 1-(4-(6,7-Dimethoxy-3,4-dihydroisoquinolin-2(1H)-yl)butyl)-3-methyl-1H-benzo[d]imidazol-2 | 1121931-70-1 | 99.9% | C23H29N3O3 | 50 MG
SDP
Catalog No. 5000354870
Encompass_Preferred
CM398 is a highly selective, orally active sigma-2 receptor ligand with a Ki of 0.43 nM. It demonstrates a high sigma-1/sigma-2 selectivity ratio (1000-fold).
  • Exhibits notable affinity for dopamine (Ki=32.90 nM) and serotonin transporters (Ki=244.2 nM).
  • Shows promising anti-inflammatory analgesic effects in the formalin model of inflammatory pain in mice.
Catalog No. 50-003-54870 Supplier Medchemexpress LLC Supplier No. HY14562850MG
CM398 is a highly selective, orally active sigma-2 receptor ligand with a Ki of 0.43 nM. It demonstrates a high sigma-1/sigma-2 selectivity ratio (1000-fold).
  • Exhibits notable affinity for dopamine (Ki=32.90 nM) and serotonin transporters (Ki=244.2 nM).
  • Shows promising anti-inflammatory analgesic effects in the formalin model of inflammatory pain in mice.