Learn More
Medchemexpress LLC 1-(4-(6,7-Dimethoxy-3,4-dihydroisoquinolin-2(1h)-yl)butyl)-3-methyl-1h-benzo[d]imidazol-2(3h)-one | 1121931-70-1 | 99.9% | C23H29N3O3 | 100 MG
SDP
Catalog No. 5000355325
Encompass_Preferred
CM398 is a highly selective, orally active sigma-2 receptor ligand. It exhibits a high sigma-1/sigma-2 selectivity ratio and shows notable affinity for dopamine and serotonin transporters. This compound has demonstrated promising anti-inflammatory analgesic effects in preclinical models of inflammatory pain.
  • Highly selective sigma-2 receptor ligand
  • Orally active
  • High sigma-1/sigma-2 selectivity ratio
  • Notable affinity for dopamine transporters
  • Notable affinity for serotonin transporters
  • Promising anti-inflammatory analgesic effects
Catalog No. 50-003-55325 Supplier Medchemexpress LLC Supplier No. HY145628100MG
CM398 is a highly selective, orally active sigma-2 receptor ligand. It exhibits a high sigma-1/sigma-2 selectivity ratio and shows notable affinity for dopamine and serotonin transporters. This compound has demonstrated promising anti-inflammatory analgesic effects in preclinical models of inflammatory pain.
  • Highly selective sigma-2 receptor ligand
  • Orally active
  • High sigma-1/sigma-2 selectivity ratio
  • Notable affinity for dopamine transporters
  • Notable affinity for serotonin transporters
  • Promising anti-inflammatory analgesic effects