Learn More
Medchemexpress LLC 1-(4-(6,7-Dimethoxy-3,4-dihydroisoquinolin-2(1H)-yl)butyl)-3-methyl-1H-benzo[d]imidazol-2(3H)-one | 1121931-70-1 | 99.9% | C23H29N3O3 | 10MG
SDP
Catalog No. 5000258307
Encompass_Preferred
CM398 is a selective, orally active sigma-2 receptor ligand used for receptor pharmacology and in vivo pain models. It has been characterized for receptor affinity and shows anti-inflammatory analgesic effects in preclinical studies.
  • High potency and selectivity for sigma-2 receptor (Ki = 0.43 nM).
  • Demonstrated efficacy in the formalin inflammatory pain model.
  • High purity suitable for research applications (99.85%).
  • Soluble in DMSO (100 mg/mL); supports multiple in vivo formulation options.
  • Stable under recommended storage conditions for long-term use.
Catalog No. 50-002-58307 Supplier Medchemexpress LLC Supplier No. HY14562810MG
CM398 is a selective, orally active sigma-2 receptor ligand used for receptor pharmacology and in vivo pain models. It has been characterized for receptor affinity and shows anti-inflammatory analgesic effects in preclinical studies.
  • High potency and selectivity for sigma-2 receptor (Ki = 0.43 nM).
  • Demonstrated efficacy in the formalin inflammatory pain model.
  • High purity suitable for research applications (99.85%).
  • Soluble in DMSO (100 mg/mL); supports multiple in vivo formulation options.
  • Stable under recommended storage conditions for long-term use.