Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-[4-(6-bromo-1,3-benzodioxol-5-yl)-3a,4,5,9b-tetrahydro-3H-cyclopenta[c]quinolin-8-yl]ethanone | 881639-98-1 | MFCD16618392 | 99.8% | 412.28 | C21H18BrNO3 | 100 MG
SDP

Catalog No. 5000239727
Encompass_Preferred

G-1 is a nonsteroidal, high-affinity, selective agonist of the GPR30 (GPER) receptor used as a research reagent to probe estrogen-related signaling. The compound is supplied as a powdered research chemical with reported molecular weight 412.28 and CAS number 881639-98-1; solubility and storage recommendations are provided for laboratory handling.

  • High-affinity GPR30 (GPER) agonist with Ki = 11 nM.
  • Supplied as a stable powder for laboratory use.
  • Soluble in DMSO at 50 mg/mL with ultrasonic assistance.
  • Recommended storage conditions for powder and solutions are provided.
  • Useful for estrogen-related signaling and receptor pharmacology studies.

Catalog No. 50-002-39727 Supplier Medchemexpress LLC Supplier No. HY107216100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

G-1 is a nonsteroidal, high-affinity, selective agonist of the GPR30 (GPER) receptor used as a research reagent to probe estrogen-related signaling. The compound is supplied as a powdered research chemical with reported molecular weight 412.28 and CAS number 881639-98-1; solubility and storage recommendations are provided for laboratory handling.

  • High-affinity GPR30 (GPER) agonist with Ki = 11 nM.
  • Supplied as a stable powder for laboratory use.
  • Soluble in DMSO at 50 mg/mL with ultrasonic assistance.
  • Recommended storage conditions for powder and solutions are provided.
  • Useful for estrogen-related signaling and receptor pharmacology studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.