Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-[4-(6-bromo-1,3-benzodioxol-5-yl)-3a,4,5,9b-tetrahydrocyclopenta[c]quinolin-8-yl]ethanone | 881639-98-1 | 99.8% | 412.28 g·mol⁻¹ | C21H18BrNO3 | 50 MG
SDP

Catalog No. 5000239967
Encompass_Preferred

G-1 is a nonsteroidal, high-affinity, selective agonist of GPR30 (G protein-coupled estrogen receptor) with a reported Ki of 11 nM. It is supplied as a research reagent with formula C21H18BrNO3, molecular weight 412.28 g·mol⁻¹, and high reported purity suitable for in vitro signaling studies.

  • Selective GPR30 agonist (Ki = 11 nM).
  • High purity (99.76%).
  • Molecular formula C21H18BrNO3 and molecular weight 412.28 g·mol⁻¹.
  • Soluble in DMSO for assay preparation.
  • Intended for research use only; not for human use.

Catalog No. 50-002-39967 Supplier Medchemexpress LLC Supplier No. HY10721650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

G-1 is a nonsteroidal, high-affinity, selective agonist of GPR30 (G protein-coupled estrogen receptor) with a reported Ki of 11 nM. It is supplied as a research reagent with formula C21H18BrNO3, molecular weight 412.28 g·mol⁻¹, and high reported purity suitable for in vitro signaling studies.

  • Selective GPR30 agonist (Ki = 11 nM).
  • High purity (99.76%).
  • Molecular formula C21H18BrNO3 and molecular weight 412.28 g·mol⁻¹.
  • Soluble in DMSO for assay preparation.
  • Intended for research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.