Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4,7,10,13,16-Hexaazacyclooctadecane Hexahydrochloride 98.0+%, TCI America™
SDP

Catalog No. H1215100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1215100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H1215100MG Supplier TCI America Supplier No. H1215100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 58105-91-2
Molecular Formula C12H36Cl6N6
Molecular Weight (g/mol) 477.162
InChI Key GYXOJYBVPYIMAW-UHFFFAOYSA-N
Synonym Hexacyclen Hexahydrochloride
PubChem CID 12710210
IUPAC Name 1,4,7,10,13,16-hexazacyclooctadecane;hexahydrochloride
SMILES C1CNCCNCCNCCNCCNCCN1.Cl.Cl.Cl.Cl.Cl.Cl

Specifications

Color White
Quantity 100 mg
Formula Weight 477.16
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,4,7,10,13,16-Hexaazacyclooctadecane Hexahydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.