Learn More
Medchemexpress LLC 1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid, α1-[3-[(2-aminoethyl)amino]-3-oxopropyl]- | 1639843-65-4 | 100.0% | 518.56 g/mol | C21H38N6O9 | 5 MG
SDP
Catalog No. 5000201935
Encompass_Preferred
NH2 DOTA-GA is a DOTA-based bifunctional metal chelator used to prepare radionuclide drug conjugates and to complex metal ions such as Gd(III) for radiolabeling and imaging research applications.
  • Bifunctional chelator for conjugation and radiolabeling.
  • Forms stable complexes with metal ions such as Gd(III).
  • High reported purity suitable for research use.
  • White to off-white solid with cold storage recommended for long-term stability.
  • Used in synthesis of radionuclide drug conjugates and contrast agents.
Catalog No. 50-002-01935 Supplier Medchemexpress LLC Supplier No. HY1645695MG
NH2 DOTA-GA is a DOTA-based bifunctional metal chelator used to prepare radionuclide drug conjugates and to complex metal ions such as Gd(III) for radiolabeling and imaging research applications.
  • Bifunctional chelator for conjugation and radiolabeling.
  • Forms stable complexes with metal ions such as Gd(III).
  • High reported purity suitable for research use.
  • White to off-white solid with cold storage recommended for long-term stability.
  • Used in synthesis of radionuclide drug conjugates and contrast agents.