Learn More
Medchemexpress LLC 1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetic acid, α-(4-isothiocyanatobenzyl)amino derivative | 2194580-71-5 | 99.2% | 622.69 | C27H38N6O9S | 10 MG
SDP
Catalog No. 5000287955
Encompass_Preferred
p-NCS-Bz-DOTA-GA is a DOTA-based bifunctional chelator featuring a para-isothiocyanatobenzyl reactive group for covalent conjugation and a DOTA-GA core for metal coordination. It is intended for preparing radiolabeled bioconjugates and metal-chelating drug conjugates, and is supplied in both solid form and as a 10 mM solution in DMSO.
  • Contains a para-isothiocyanate reactive handle for amine coupling.
  • DOTA-GA chelator core provides strong metal coordination.
  • Suitable for radiolabeling peptides, proteins, and antibodies.
  • Available as solid and as a ready-to-use 10 mM solution in DMSO.
  • High purity for research applications.
  • White to off-white solid; handle under anhydrous conditions to preserve stability.
Catalog No. 50-002-87955 Supplier Medchemexpress LLC Supplier No. HY15918110MG
p-NCS-Bz-DOTA-GA is a DOTA-based bifunctional chelator featuring a para-isothiocyanatobenzyl reactive group for covalent conjugation and a DOTA-GA core for metal coordination. It is intended for preparing radiolabeled bioconjugates and metal-chelating drug conjugates, and is supplied in both solid form and as a 10 mM solution in DMSO.
  • Contains a para-isothiocyanate reactive handle for amine coupling.
  • DOTA-GA chelator core provides strong metal coordination.
  • Suitable for radiolabeling peptides, proteins, and antibodies.
  • Available as solid and as a ready-to-use 10 mM solution in DMSO.
  • High purity for research applications.
  • White to off-white solid; handle under anhydrous conditions to preserve stability.