Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,4,7,10-Tetraazacyclododecane-1,4,7,10-Tetraacetic Acid

Catalog No. 86734250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mg
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
86-734-250MG 50 mg
86-734-50MG 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 86-734-250MG Supplier Sigma Aldrich Supplier No. 86734250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid (DOTA) is a macrocyclic complexing agent.It has been used for radiolabeling of carbon nanotube bioconjugates by chelating 64Cu radioisotope.DOTA can be activated with N-hydroxysulfosuccinimidyl for conjugation with monoclonal antibodies in clinical radioimmunotherapy.It can form complexes with gadolinium for application as MRI contrast agents.Metal complexes of DOTA-peptide conjugates can be used as therapeutic radiopharmaceuticals.

Specifications

CAS 60239-18-1
Molecular Formula C16H28N4O8 -+ xH2O
Linear Formula C16H28N4O8 -+ xH2O
MDL Number MFCD00068657
Quantity 50 mg
Synonym DOTA
Molecular Weight (g/mol) 404.42 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.