Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,4,7,10-Tetraazacyclododecane-1,4,7,10-Tetraacetic Acid

Catalog No. 8673450MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mg
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
86-734-50MG 50 g
86-734-250MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 86-734-50MG Supplier Sigma Aldrich Supplier No. 8673450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid (DOTA) is a macrocyclic complexing agent.It has been used for radiolabeling of carbon nanotube bioconjugates by chelating 64Cu radioisotope.DOTA can be activated with N-hydroxysulfosuccinimidyl for conjugation with monoclonal antibodies in clinical radioimmunotherapy.It can form complexes with gadolinium for application as MRI contrast agents.Metal complexes of DOTA-peptide conjugates can be used as therapeutic radiopharmaceuticals.

Specifications

CAS 5574-97-0
Melting Point 151°C to 154°C
Molecular Formula C16H38NO4P
Linear Formula (CH3CH2CH2CH2)4N[OP(OH)2O]
MDL Number MFCD00064526
Quantity 50 g
Molecular Weight (g/mol) 339.45 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.