Learn More
Medchemexpress LLC 1,4,7,10-tetraazacyclododecane-1,4,7-tris(acetic acid)-10-(3-butynylacetamide) | 2125661-62-1 | MFCD35160514 | 97.0% | 455.51 | C20H33N5O7 | 5mg
SDP
Catalog No. 5000269443
Encompass_Preferred
Butyne-DOTAis a bifunctional chelator (Bifunctional Chelator BFC) and a macrocyclic DOTA derivative used for tumor pre-targeting Butyne-DOTA can be used for conjugation of peptides and radionuclides
Catalog No. 50-002-69443 Supplier Medchemexpress LLC Supplier No. HYW7820785MG
Butyne-DOTAis a bifunctional chelator (Bifunctional Chelator BFC) and a macrocyclic DOTA derivative used for tumor pre-targeting Butyne-DOTA can be used for conjugation of peptides and radionuclides