Learn More
Medchemexpress LLC 1,4,7,10-tetraazacyclododecane-1,7-diacetic acid, 1,7-bis(1,1-dimethylethyl) ester | 162148-48-3 | MFCD09263303 | 98.0% | 400.56 g·mol⁻¹ | C20H40N4O4 | 5 G
SDP
Catalog No. 5000280594
Encompass_Preferred
DO2A-tert-butyl ester is a tert-butyl-protected DOTA-like bifunctional chelator used as a synthetic building block for preparing metal-chelator conjugates and radionuclide-labeled probes in biochemical and radiochemistry applications. The compound has molecular formula C20H40N4O4 and a molecular weight of 400.56 g·mol⁻¹; store protected from light at 4°C and follow solvent storage guidelines for long-term stability.
  • Suitable for chelator-biomolecule conjugation
  • Protected ester functionality for controlled deprotection
  • Applicable for radionuclide labeling workflows
  • Solid reagent with defined molecular composition
  • Stable under recommended storage conditions
Catalog No. 50-002-80594 Supplier Medchemexpress LLC Supplier No. HYW2835565G
DO2A-tert-butyl ester is a tert-butyl-protected DOTA-like bifunctional chelator used as a synthetic building block for preparing metal-chelator conjugates and radionuclide-labeled probes in biochemical and radiochemistry applications. The compound has molecular formula C20H40N4O4 and a molecular weight of 400.56 g·mol⁻¹; store protected from light at 4°C and follow solvent storage guidelines for long-term stability.
  • Suitable for chelator-biomolecule conjugation
  • Protected ester functionality for controlled deprotection
  • Applicable for radionuclide labeling workflows
  • Solid reagent with defined molecular composition
  • Stable under recommended storage conditions