Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,4,7,10-tetraoxacyclododecane | 294-93-9 | MFCD00005103 | 98.0% | 176.21 g/mol | C8H16O4 | 1 G
SDP

Catalog No. 5000303779
Encompass_Preferred

12-Crown-4 is a crown ether used as a phase-transfer catalyst and as a complexing agent for lithium ions in laboratory research. It promotes selective binding of small alkali metal cations and is used in synthetic, catalytic, and analytical workflows. Supplied for research use only.

  • Effective phase-transfer catalyst for lithium ions.
  • Selective complexation of small alkali metal cations.
  • High purity (98.0%).
  • Molecular weight 176.21 g/mol; formula C8H16O4.
  • Available in small laboratory pack sizes suitable for research (for example, 1 g).
  • CAS number 294-93-9 for unambiguous identification.

Catalog No. 50-003-03779 Supplier Medchemexpress LLC Supplier No. HYW0166931G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

12-Crown-4 is a crown ether used as a phase-transfer catalyst and as a complexing agent for lithium ions in laboratory research. It promotes selective binding of small alkali metal cations and is used in synthetic, catalytic, and analytical workflows. Supplied for research use only.

  • Effective phase-transfer catalyst for lithium ions.
  • Selective complexation of small alkali metal cations.
  • High purity (98.0%).
  • Molecular weight 176.21 g/mol; formula C8H16O4.
  • Available in small laboratory pack sizes suitable for research (for example, 1 g).
  • CAS number 294-93-9 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.