Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4,7-Triazacyclononane 98.0+%, TCI America™
SDP

Catalog No. T18785G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18785G 5 g
T1878200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18785G Supplier TCI America Supplier No. T18785G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4730-54-5
Molecular Formula C6H15N3
Molecular Weight (g/mol) 129.207
InChI Key ITWBWJFEJCHKSN-UHFFFAOYSA-N
Synonym Octahydro-1H-1,4,7-triazonine
PubChem CID 188318
ChEBI CHEBI:37405
IUPAC Name 1,4,7-triazonane
SMILES C1CNCCNCCN1

Specifications

Melting Point 45°C
Color White-Yellow
Boiling Point 113°C
Quantity 5 g
UN Number 3259
Formula Weight 129.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4,7-Triazacyclononane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.