Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,4,7-trimethyl-1,4,7-triazacyclononane | 96556-05-7 | MFCD00006093 | 98.5% | 171.28 g/mol | C9H21N3 | 5 G
SDP

Catalog No. 5000209929
Encompass_Preferred

1,4,7-trimethyl-1,4,7-triazacyclononane is a small cyclic tertiary amine used as a reagent in biochemical assays and as a crosslinking or reactive reagent for research applications.

  • used as a reagent in biochemical assays
  • serves as a crosslinking or reactive reagent in research
  • molecular formula c9h21n3
  • molecular weight 171.28 g/mol
  • high purity, typically about 98.5%

Catalog No. 50-002-09929 Supplier Medchemexpress LLC Supplier No. HYW0513915G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,4,7-trimethyl-1,4,7-triazacyclononane is a small cyclic tertiary amine used as a reagent in biochemical assays and as a crosslinking or reactive reagent for research applications.

  • used as a reagent in biochemical assays
  • serves as a crosslinking or reactive reagent in research
  • molecular formula c9h21n3
  • molecular weight 171.28 g/mol
  • high purity, typically about 98.5%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.