Learn More
Strem, An Ascensus Company CAS# 4741-99-5. 1g. 1,4,8,11-Tetraazaundecane, min. 97%. MFCD00008174
SDP
Catalog No. 502045219
Encompass
CAS# 4741-99-5. 1g. 1,4,8,11-Tetraazaundecane, min. 97%. MFCD00008174. Molecular Weight: 160.26. Molecular Formula: C7H20N4. Color/form: colorless to pale yellow liq. Strem# 07-1965. http://www.strem.com/catalog/v/07-1965/
Catalog No. 50-204-5219 Supplier Strem, An Ascensus Company Supplier No. 0719651G
CAS# 4741-99-5. 1g. 1,4,8,11-Tetraazaundecane, min. 97%. MFCD00008174. Molecular Weight: 160.26. Molecular Formula: C7H20N4. Color/form: colorless to pale yellow liq. Strem# 07-1965. http://www.strem.com/catalog/v/07-1965/