Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Acetylpiperidino)ethan-1-one, 97%, Thermo Scientific™

Catalog No. CD11879DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CD11879DE 5 g Amber Glass Bottle
CD11879CB 250 mg Amber Glass Bottle
CD11879DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CD11879DE Supplier Thermo Scientific Chemicals Supplier No. CD11879DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 162368-01-6
Molecular Formula C9H15NO2
Molecular Weight (g/mol) 169.22
MDL Number MFCD00106774
InChI Key VFZXZEJGLKOTBW-UHFFFAOYSA-N
Synonym 1-4-acetylpiperidino ethan-1-one, 1-1-acetylpiperidin-4-yl ethanone, 1,4-diacetylpiperidine, 1-4-acetylpiperidin-1-yl ethan-1-one, 1,1'-piperidine-1,4-diyl diethanone, 1-1-acetyl-piperidin-4-yl-ethanone, piperidine, 1,4-diacetyl-9ci, 1-4-acetylpiperidin-1-yl ethanone, ethanone,1-1-acetyl-4-piperidinyl, piperidine, 1,4-diacetyl
PubChem CID 2777250
IUPAC Name 1-(1-acetylpiperidin-4-yl)ethanone
SMILES CC(=O)C1CCN(CC1)C(C)=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 169.22
Percent Purity 97%
Chemical Name or Material 1-(4-Acetylpiperidino)ethan-1-one
Hazard Category Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.