Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Amino-2-hydroxyphenyl)ethan-1-one, Tech., Thermo Scientific™

Catalog No. BTB10056EA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB10056EA 10 g Amber Glass Bottle
BTB10056DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB10056EA Supplier Thermo Scientific Chemicals Supplier No. BTB10056EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2476-29-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00100636
InChI Key QQZFVONVJPXCSQ-UHFFFAOYSA-N
Synonym 1-4-amino-2-hydroxyphenyl ethanone, 1-4-amino-2-hydroxyphenyl ethan-1-one, 4'-amino-2'-hydroxyacetophenone, 1-4-amino-2-hydroxyphenyl ethane-1-one, ethanone, 1-4-amino-2-hydroxyphenyl, 1-4-amino-2-hydroxy-phenyl ethanone, 2-acetyl-5-aminophenol, intermediates-zcf02667, amino-2-hydroxyphenyl ethanone, 4-acetyl-3-hydroxyaniline
PubChem CID 459296
IUPAC Name 1-(4-amino-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=C(O)C=C(N)C=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 151.16
Percent Purity Tech.
Grade Technical
Chemical Name or Material 1-(4-Amino-2-hydroxyphenyl)ethan-1-one
Hazard Category Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.