Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Aminobenzyl)-1H-1,2,4-triazole, 98%, Thermo Scientific™

Catalog No. AAH6662014
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6662014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6662014 Supplier Thermo Scientific Chemicals Supplier No. H6662014

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119192-10-8
Molecular Formula C9H10N4
Molecular Weight (g/mol) 174.207
MDL Number MFCD00973306
InChI Key ZGLQVRIVLWGDNA-UHFFFAOYSA-N
Synonym 4-1h-1,2,4-triazol-1-ylmethyl aniline, 4-1h-1,2,4-triazol-1-yl methyl aniline, 1-4-aminobenzyl-1,2,4-triazole, 4-1,2,4-triazol-1-ylmethyl aniline, 4-1h-1,2,4-triazol-1-yl-methyl benzenamine, 4-1,2,4 triazol-1-ylmethylphenylamine, benzenamine, 4-1h-1,2,4-triazol-1-ylmethyl, 4-1,2,4 triazol-1-ylmethyl-phenylamine, 4-1h-1,2,4-triazol-1-yl-methyl aniline, 4-1h-1,2,4-triazol-1-ylmethyl benzenamine
PubChem CID 821219
IUPAC Name 4-(1,2,4-triazol-1-ylmethyl)aniline
SMILES C1=CC(=CC=C1CN2C=NC=N2)N

Specifications

Melting Point 126°C to 130°C
Color White to Yellow
Quantity 25 g
UN Number UN2811
Beilstein 5934828
Formula Weight 174.21
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-(4-Aminobenzyl)-1H-1,2,4-triazole
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.