Learn More
Medchemexpress LLC 1-(4-(aminomethyl)benzyl)-2-butyl-1H-imidazo[4,5-c]quinolin-4-amine dihydrochloride | 1620278-72-9 | MFCD31657341 | 99.7% | 432.39 g/mol | C22H27Cl2N5 | 1 ML
SDP
Catalog No. 5000243548
Encompass_Preferred
TLR7/8 agonist 1 dihydrochloride is a dual-agonistic imidazoquinoline research compound that activates toll-like receptors 7 and 8 to induce innate immune responses. It is used in cell-based assays and in vivo studies to stimulate immune signaling and can serve as a derivatizable precursor for covalent labeling while retaining biological activity.
  • Dual agonist of toll-like receptors 7 and 8, inducing innate immune signaling.
  • Low-nanomolar agonistic potency (≈50 nM for TLR7, ≈55 nM for TLR8).
  • Available as a DMSO solution for convenient dosing in cell assays.
  • Suitable for use in cell-based and in vivo immunostimulatory studies.
  • Precursor amenable to covalent modification without significant loss of activity.
  • Stable when stored sealed at 4°C; in solution recommended -80°C for long-term storage.
Catalog No. 50-002-43548 Supplier Medchemexpress LLC Supplier No. HY103698A10MM 1ML
TLR7/8 agonist 1 dihydrochloride is a dual-agonistic imidazoquinoline research compound that activates toll-like receptors 7 and 8 to induce innate immune responses. It is used in cell-based assays and in vivo studies to stimulate immune signaling and can serve as a derivatizable precursor for covalent labeling while retaining biological activity.
  • Dual agonist of toll-like receptors 7 and 8, inducing innate immune signaling.
  • Low-nanomolar agonistic potency (≈50 nM for TLR7, ≈55 nM for TLR8).
  • Available as a DMSO solution for convenient dosing in cell assays.
  • Suitable for use in cell-based and in vivo immunostimulatory studies.
  • Precursor amenable to covalent modification without significant loss of activity.
  • Stable when stored sealed at 4°C; in solution recommended -80°C for long-term storage.