Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(4-aminomethyl-phenyl)-pyrrolidin-2-one hydrochloride

Catalog No. CDS013032 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS013032 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS013032 Supplier Sigma Aldrich Supplier No. CDS01303225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,3,5-tris(trifluoromethyl)benzene is a tri-substituted benzene
  • It has been synthesized by reacting benzene-1,3,5-tricarboxylic acid with sf4
  • Reports suggest that 1,3,5-tris(trifluoromethyl)benzene is a nonanesthetics and shows convulsant properties
  • Its vapor absorption spectra and corresponding solution spectra has been investigated
  • Its reaction with methyl lithium has been analyzed using semiempirical reaction modelling.1,3,5-tris(trifluoromethyl)benzene was used as a starting material in the synthesis of bis(2,4,6-tris(trifluoromethyl)phenyl)chloropnictines.it may be used as a starting material in the synthesis of 2,4,6-tris(trifluoromethyl)benzoic acid by reacting with n-butyllithium and carbon dioxide and in the synthesis of lithio derivative, via direct metalation with n-butyllithium

Specifications

Molecular Formula C11H15ClN2O
Linear Formula C11H15ClN2O
MDL Number MFCD06799747
Quantity 5 g
Molecular Weight (g/mol) 226.7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.