Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Anhydroerythritol 98.0+%, TCI America™
SDP

Catalog No. A18395G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A18395G Supplier TCI America Supplier No. A18395G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4358-64-9
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.105
MDL Number MFCD00075210
InChI Key SSYDTHANSGMJTP-ZXZARUISSA-N
Synonym cis-3,4-Tetrahydrofurandiol
PubChem CID 641773
IUPAC Name (3S,4R)-oxolane-3,4-diol
SMILES C1C(C(CO1)O)O

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
Formula Weight 104.11
Percent Purity ≥98.0% (GC)
Chemical Name or Material 1,4-Anhydroerythritol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.