Learn More
MSC Value Collection Ball End Mill, 1/4 in, 1 ea
Catalog No. NC1192843
Encompass
A high-speed steel ball end mill designed for precision milling applications. This single-end tool features a right-hand cut and spiral flutes for efficient material removal.
  • Mill diameter: 1/4 in
  • Length of cut: 1-1/4 in
  • Four flutes
  • High speed steel construction
  • Bright/uncoated finish
  • Centercutting capability
Catalog No. NC1192843 Supplier MSC Supplier No. 01892165
A high-speed steel ball end mill designed for precision milling applications. This single-end tool features a right-hand cut and spiral flutes for efficient material removal.
  • Mill diameter: 1/4 in
  • Length of cut: 1-1/4 in
  • Four flutes
  • High speed steel construction
  • Bright/uncoated finish
  • Centercutting capability