Learn More
Medchemexpress LLC 1,4-Benzenediamine, N1-(1,4-dimethylpentyl)-N4-phenyl- | 3081-01-4 | 98.3% | 282.42 | 10 G
SDP
Catalog No. 5000386997
Encompass_Preferred
7-PPD (DMPPD) is a rubber antioxidant that scavenges free radicals and inhibits the oxidative degradation of rubber chains. It shows promise for research in tire manufacturing and rubber product processing. It is for research use only and not sold to patients.
  • Acts as a rubber antioxidant
  • Scavenges free radicals
  • Inhibits oxidative degradation of rubber chains
  • Shows potential for research in tire manufacturing
  • Shows potential for research in rubber product processing
Catalog No. 50-003-86997 Supplier Medchemexpress LLC Supplier No. HY17674710G
7-PPD (DMPPD) is a rubber antioxidant that scavenges free radicals and inhibits the oxidative degradation of rubber chains. It shows promise for research in tire manufacturing and rubber product processing. It is for research use only and not sold to patients.
  • Acts as a rubber antioxidant
  • Scavenges free radicals
  • Inhibits oxidative degradation of rubber chains
  • Shows potential for research in tire manufacturing
  • Shows potential for research in rubber product processing