Learn More
1,4-Benzenediboronic acid, 96%
Description
It is used for palladium-catalyzed sequential alkenylation and conjugate addition reactions, scholl cyclizations, Suzuki-Miyaura cross-coupling reactions. It is also used in the preparation of indolizine derivatives as OLEDs, in organic thin-film transistors, Fluorescence and solution-processable coordination polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used for palladium-catalyzed sequential alkenylation and conjugate addition reactions, scholl cyclizations, Suzuki-Miyaura cross-coupling reactions. It is also used in the preparation of indolizine derivatives as OLEDs, in organic thin-film transistors, Fluorescence and solution-processable coordination polymers.
Solubility
Soluble in water 2.5%.
Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 5 g |
| Beilstein | 2836921 |
| Solubility Information | Soluble in water 2.5%. |
| Formula Weight | 165.75 |
| Percent Purity | 96% |
| Chemical Name or Material | 1,4-Benzenediboronic acid |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H301 Toxic if swallowed. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.