Learn More
Medchemexpress LLC 1,4-Benzenedicarboxylic acid | 100-21-0 | 99.9% | 166.13 | 10 G
SDP
Catalog No. 5000452513
Encompass_Preferred
Terephthalic acid is an isomer and a precursor of polyester PET, which can be used to make clothing and plastics. It can also be utilized to build skin tumor models.
  • Can be used in animal modeling to create skin tumor models.
  • It is metabolized in the body to form calcium terephthalate or calcium hydrogen terephthalate, which can lead to bladder stone formation.
  • The lithogenic effect is more pronounced in weanling mice at higher doses.
  • Promotes excessive cell proliferation and induces skin tumor formation by disrupting the p16-cyclin D1/Cdk4-Rb cell cycle regulatory pathway.
  • Specific modeling methods for skin tumor induction involve topical application in C57BL/6 mice.
Catalog No. 50-004-52513 Supplier Medchemexpress LLC Supplier No. HYW01009810G
Terephthalic acid is an isomer and a precursor of polyester PET, which can be used to make clothing and plastics. It can also be utilized to build skin tumor models.
  • Can be used in animal modeling to create skin tumor models.
  • It is metabolized in the body to form calcium terephthalate or calcium hydrogen terephthalate, which can lead to bladder stone formation.
  • The lithogenic effect is more pronounced in weanling mice at higher doses.
  • Promotes excessive cell proliferation and induces skin tumor formation by disrupting the p16-cyclin D1/Cdk4-Rb cell cycle regulatory pathway.
  • Specific modeling methods for skin tumor induction involve topical application in C57BL/6 mice.