Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,4-Benzenedimethanol

Catalog No. B300050GA Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B300050GA 50 g
B300010GA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B300050GA Supplier Sigma Aldrich Supplier No. B300050GA
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,4-Benzenedimethanol

Specifications

CAS 589-29-7
Melting Point 114°C to 118°C (lit.)
Boiling Point 138°C to 143°C (1 mmHg)
Molecular Formula C8H10O2
Linear Formula C6H4(CH2OH)2
MDL Number MFCD00004665
Quantity 50 g
Synonym p-Phenylene dicarbinol; p-Phenylenedimethanol; p-Xylene-alpha,alpha-diol; p-Xylylene dialcohol; NSC 5097
Molecular Weight (g/mol) 138.16
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.