Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,4-Benzenediol, 2-(hydroxymethyl)- | 495-08-9 | MFCD06202616 | 98.9% | 140.14 g/mol | C7H8O3 | 100 MG
SDP

Catalog No. 5000281348
Encompass_Preferred

Gentisyl alcohol is a small-molecule research chemical reported to have anti-Staphylococcus aureus activity. It is supplied as a high-purity solid for antibacterial research and in vitro assays, with documented solubility and storage recommendations to preserve stability.

  • Purity 98.9%
  • CAS number 495-08-9
  • Chemical formula C7H8O3; molecular weight 140.14 g/mol
  • Solubility DMSO 100 mg/mL (may require ultrasonic)
  • Storage -20°C under nitrogen; in solvent -80°C (6 months) or -20°C (1 month)
  • Intended use antibacterial research and in vitro studies
  • Available quantity 100 mg

Catalog No. 50-002-81348 Supplier Medchemexpress LLC Supplier No. HYW248907100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gentisyl alcohol is a small-molecule research chemical reported to have anti-Staphylococcus aureus activity. It is supplied as a high-purity solid for antibacterial research and in vitro assays, with documented solubility and storage recommendations to preserve stability.

  • Purity 98.9%
  • CAS number 495-08-9
  • Chemical formula C7H8O3; molecular weight 140.14 g/mol
  • Solubility DMSO 100 mg/mL (may require ultrasonic)
  • Storage -20°C under nitrogen; in solvent -80°C (6 months) or -20°C (1 month)
  • Intended use antibacterial research and in vitro studies
  • Available quantity 100 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.