Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,4-Benzodioxane-6-boronic acid | 164014-95-3 | 100.0% | 179.97 | 10 G
SDP

Catalog No. 5000404336
Encompass_Preferred

1,4-Benzodioxane-6-boronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • Molecular weight: 179.97
  • Formula: C8H9BO4
  • CAS number: 164014-95-3
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 99.97%
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder: -20°C for 3 years; 4°C for 2 years. In solvent: -80°C for 6 months; -20°C for 1 month

Catalog No. 50-004-04336 Supplier Medchemexpress LLC Supplier No. HY3451110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,4-Benzodioxane-6-boronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • Molecular weight: 179.97
  • Formula: C8H9BO4
  • CAS number: 164014-95-3
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 99.97%
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder: -20°C for 3 years; 4°C for 2 years. In solvent: -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.