Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,4-Benzodioxane-6-Boronic Acid

Catalog No. 6359955G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-599-55G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 63-599-55G Supplier Sigma Aldrich Supplier No. 6359955G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant involved in:Addition reactions with 1,8-naphthyridine N-oxidesIodocyclization and palladium-catalyzed coupling reactions for synthesis of pyranoquinolinesSuzuki coupling reactions for synthesis of combretastatin analogsSuzuki-Miyaura coupling reactions for synthesis of aryl-substituted oxabenzindoles and methanobenzindolesPalladium-catalyzed oxyarylation of Heck reaction intermediatesC-H functionalization of quinonesContains varying amounts of anhydride

Specifications

CAS 78887-39-5
Melting Point 135°C
Molecular Formula C8H10BNO3
Linear Formula CH3CONHC6H4B(OH)2
MDL Number MFCD00236013
Quantity 1 g
Synonym 6-(1,4-Benzodioxanyl)boronic acid; 1,4-Benzodioxan-6-boronic acid
Molecular Weight (g/mol) 178.98 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.