Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,4-Benzodioxane-6-carboxylic acid | 4442-54-0 | 180.16 | 5 G
SDP

Catalog No. 5000448999
Encompass_Preferred

2,3-Dihydro-1,4-benzodioxine-6-carboxylic acid is a biochemical reagent used as a biological material or organic compound for life science research. It is intended for research use only.

  • Appearance: solid, white to off-white
  • Molecular weight: 180.16
  • Purity: 99.9%
  • Solubility (in vitro): DMSO: ≥ 100 mg/mL
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Applications: biochemical assay reagents, biological material, organic compound, inhibitor studies

Catalog No. 50-004-48999 Supplier Medchemexpress LLC Supplier No. HYW0026235G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,3-Dihydro-1,4-benzodioxine-6-carboxylic acid is a biochemical reagent used as a biological material or organic compound for life science research. It is intended for research use only.

  • Appearance: solid, white to off-white
  • Molecular weight: 180.16
  • Purity: 99.9%
  • Solubility (in vitro): DMSO: ≥ 100 mg/mL
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Applications: biochemical assay reagents, biological material, organic compound, inhibitor studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.