Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,4-Benzodioxane-6-carboxylic acid | 4442-54-0 | MFCD00463509 | 180.16 | 10 G
SDP

Catalog No. 5000449509
Encompass_Preferred

2,3-Dihydro-1,4-benzodioxine-6-carboxylic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • CAS number: 4442-54-0
  • Molecular formula: C9H8O4
  • Molecular weight: 180.16
  • Purity: 99.96%
  • Appearance: Solid, white to off-white
  • Storage: Powder -20°C 3 years, 4°C 2 years
  • Storage in solvent: -80°C 6 months, -20°C 1 month

Catalog No. 50-004-49509 Supplier Medchemexpress LLC Supplier No. HYW00262310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,3-Dihydro-1,4-benzodioxine-6-carboxylic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • CAS number: 4442-54-0
  • Molecular formula: C9H8O4
  • Molecular weight: 180.16
  • Purity: 99.96%
  • Appearance: Solid, white to off-white
  • Storage: Powder -20°C 3 years, 4°C 2 years
  • Storage in solvent: -80°C 6 months, -20°C 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.