Learn More
Medchemexpress LLC 1,4-Bis(2-methylstyryl)-benzene | 13280-61-0 | ≥99.0% | 310.43 | 1 G
SDP
Catalog No. 5000453836
Encompass_Preferred
1,4-Bis(2-methylstyryl)-benzene,99% (Bis-MSB,99%) is a biochemical assay reagent intended for research use only. This compound is used in various laboratory applications.
  • Suitable for biochemical assays
  • Can be used as an enzyme substrate
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-53836 Supplier Medchemexpress LLC Supplier No. HYW0113461G
1,4-Bis(2-methylstyryl)-benzene,99% (Bis-MSB,99%) is a biochemical assay reagent intended for research use only. This compound is used in various laboratory applications.
  • Suitable for biochemical assays
  • Can be used as an enzyme substrate
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month