Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Bis(2-propynyloxy)benzene 98.0+%, TCI America™
SDP

Catalog No. B46075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46075G 5 g
B46071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46075G Supplier TCI America Supplier No. B46075G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34596-36-6
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00224313
InChI Key ODCQCPUUPWRQKV-UHFFFAOYSA-N
Synonym 1,4-Bis(propargyloxy)benzene
PubChem CID 12933255
IUPAC Name 1,4-bis(prop-2-ynoxy)benzene
SMILES C#CCOC1=CC=C(C=C1)OCC#C

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 202°C
Quantity 5 g
Formula Weight 186.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis(2-propynyloxy)benzene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.