Learn More
Medchemexpress LLC 1,4-bis[(3,4-dimethoxyphenyl)sulfonyl]-1,4-diazepane | 447410-57-3 | MFCD02585318 | 98.9% | 500.59 | C21H28N2O8S2 | 25 MG
SDP
Catalog No. 5000247734
Encompass_Preferred
SB756050 is a selective agonist of the TGR5 (GPBAR1) bile-acid receptor used for in vitro and preclinical research. It is supplied as a characterized compound with defined molecular formula C21H28N2O8S2, molecular weight 500.59, and high purity to support reproducible assay results. Intended for research use only; not for human or clinical applications.
  • Selective TGR5 (GPBAR1) agonist suitable for receptor pharmacology studies.
  • High purity for reproducible biochemical and cellular assays.
  • Available in multiple vial sizes and solution formats for flexibility.
  • Stable under recommended storage conditions for long-term use.
  • Intended for research use only; not for human use.
Catalog No. 50-002-47734 Supplier Medchemexpress LLC Supplier No. HY10201625MG
SB756050 is a selective agonist of the TGR5 (GPBAR1) bile-acid receptor used for in vitro and preclinical research. It is supplied as a characterized compound with defined molecular formula C21H28N2O8S2, molecular weight 500.59, and high purity to support reproducible assay results. Intended for research use only; not for human or clinical applications.
  • Selective TGR5 (GPBAR1) agonist suitable for receptor pharmacology studies.
  • High purity for reproducible biochemical and cellular assays.
  • Available in multiple vial sizes and solution formats for flexibility.
  • Stable under recommended storage conditions for long-term use.
  • Intended for research use only; not for human use.