Learn More
Medchemexpress LLC 1,4-bis(4,5-diphenyl-1H-imidazol-2-yl)benzene | 4051-59-6 | MFCD00231029 | 98.0% | 514.62 g·mol⁻¹ | C36H26N4 | 50 MG
SDP
Catalog No. 5000246098
Encompass_Preferred
MSX-130 is a small-molecule CXCR4 antagonist for preclinical research, reported to inhibit cancer metastasis. It is supplied as a solid with high purity and recommended for in vitro studies using DMSO preparations.
  • Targets CXCR4 and inhibits cancer metastasis.
  • High purity (98.0%) suitable for biochemical assays.
  • Soluble in DMSO at approximately 2 mg/mL with sonication and warming.
  • Powder stable at -20°C for extended storage; solutions require low-temperature storage for long-term stability.
  • Supplied as a 50 mg solid for laboratory research use.
Catalog No. 50-002-46098 Supplier Medchemexpress LLC Supplier No. HY10301050MG
MSX-130 is a small-molecule CXCR4 antagonist for preclinical research, reported to inhibit cancer metastasis. It is supplied as a solid with high purity and recommended for in vitro studies using DMSO preparations.
  • Targets CXCR4 and inhibits cancer metastasis.
  • High purity (98.0%) suitable for biochemical assays.
  • Soluble in DMSO at approximately 2 mg/mL with sonication and warming.
  • Powder stable at -20°C for extended storage; solutions require low-temperature storage for long-term stability.
  • Supplied as a 50 mg solid for laboratory research use.